Learn More
Description
Specifications
Specifications
| Antigen | Kv7.3 |
| Applications | ELISA |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Peptide ELISA Detection limit 1:16000 |
| Formulation | Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide |
| Gene Accession No. | NP_004510.1 |
| Gene Alias | BFNC2, EBN2, KQT-like 3, KV7.3, Potassium channel subunit alpha KvLQT3, potassium channel, voltage-gated, subfamily Q, member 3, potassium voltage-gated channel subfamily KQT member 3, potassium voltage-gated channel, KQT-like subfamily, member 3, Voltage-gated potassium channel subunit Kv7.3 |
| Gene Symbols | KCNQ3 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
