Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv7.4 Antibody (S43/6), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. NBP212899Z Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212899Z 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212899Z Supplier Novus Biologicals Supplier No. NBP212899AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kv7.4 Monoclonal specifically detects Kv7.4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Kv7.4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone S43/6
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen
Gene Alias DFNA2, DFNA2A, KQT-like 4, KV7.4, potassium channel KQT-like 4, Potassium channel subunit alpha KvLQT4, potassium voltage-gated channel subfamily KQT member 4, potassium voltage-gated channel, KQT-like subfamily, member 4, Voltage-gated potassium channel subunit Kv7.4
Gene Symbols KCNQ4
Host Species Mouse
Immunogen Fusion protein amino acids 2-77 of human KCNQ4
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9132
Test Specificity Detects approx 77kDa.
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.