Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv7.5 Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB051999 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB051999 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB051999 Supplier Novus Biologicals Supplier No. NBP298067AF532
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kv7.5 Polyclonal specifically detects Kv7.5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Kv7.5
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias KQT-like 5, Kv7.5, potassium channel protein, Potassium channel subunit alpha KvLQT5, potassium voltage-gated channel subfamily KQT member 5, potassium voltage-gated channel, KQT-like subfamily, member 5, Voltage-gated potassium channel subunit Kv7.5
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Kv7.5 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 56479
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.