Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv8.2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094760 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094760 100 μL
NB094761 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094760 Supplier Novus Biologicals Supplier No. NBP303519100UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kv8.2 Polyclonal specifically detects Kv8.2 in Human samples. It is validated for Western Blot.

Specifications

Antigen Kv8.2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias KV11.1, Kv8.2, MGC120515, potassium channel, subfamily V, member 2, potassium voltage-gated channel subfamily V member 2, RCD3B, voltage-gated potassium channel Kv8.2, Voltage-gated potassium channel subunit Kv8.2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 177-261 of human Kv8.2 (NP_598004.1).
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169522
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.