Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kv9.3 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB143961 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB143961 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB143961 Supplier Novus Biologicals Supplier No. NBP305922AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kv9.3 Polyclonal specifically detects Kv9.3 in Human samples. It is validated for Western Blot.

Specifications

Antigen Kv9.3
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias delayed-rectifier K(+) channel alpha subunit 3, KV9.3, MGC9481, potassium voltage-gated channel subfamily S member 3, potassium voltage-gated channel, delayed-rectifier, subfamily S, member 3, Shab-related delayed-rectifier K+ channel alpha subunit 3, voltage-gated potassium channel protein Kv9.3, voltage-gated potassium channel subunit Kv9.3
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Kv9.3 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3790
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.