Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KvBeta1 Antibody (OTI4E7), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP271344IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271344IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271344IR Supplier Novus Biologicals Supplier No. NBP271344IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

KvBeta1 Monoclonal specifically detects KvBeta1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen KvBeta1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI4E7
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AKR6A3, hKvb3, hKvBeta3, K(+) channel subunit beta-1, KCNA1Bpotassium channel beta 3 chain, Kvb1.3, KV-BETA-1, potassium channel beta3 subunit, potassium channel shaker chain beta 1a, potassium voltage-gated channel beta subunit, potassium voltage-gated channel, shaker-related subfamily, beta member 1, voltage-gated potassium channel beta-1 subunit, voltage-gated potassium channel subunit beta-1
Gene Symbols KCNAB1
Host Species Mouse
Immunogen Full length recombinant protein of human KvBeta1 (NP_751891) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7881.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.