Learn More
Description
Specifications
Specifications
| Antigen | KvBeta1 |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Clone | OTI7F12 |
| Conjugate | Alexa Fluor 647 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | AKR6A3, hKvb3, hKvBeta3, K(+) channel subunit beta-1, KCNA1Bpotassium channel beta 3 chain, Kvb1.3, KV-BETA-1, potassium channel beta3 subunit, potassium channel shaker chain beta 1a, potassium voltage-gated channel beta subunit, potassium voltage-gated channel, shaker-related subfamily, beta member 1, voltage-gated potassium channel beta-1 subunit, voltage-gated potassium channel subunit beta-1 |
| Gene Symbols | KCNAB1 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
