Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kynureninase Antibody (OTI1H1), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N271400A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N271400A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N271400A647 Supplier Novus Biologicals Supplier No. NBP271400AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Kynureninase Monoclonal specifically detects Kynureninase in Human, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen Kynureninase
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI1H1
Conjugate Alexa Fluor 647
Dilution Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 3.7.1.3, kynureninase, kynureninase (L-kynurenine hydrolase), L-kynurenine hydrolase
Gene Symbols KYNU
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-216 of human Kynureninase (NP_001028170) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 8942.0
Target Species Human, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.