Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kynurenine 3-Monooxygenase/KMO Antibody, Novus Biologicals™
SDP

Catalog No. NBP144263 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP144263 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP144263 Supplier Novus Biologicals Supplier No. NBP144263
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Kynurenine 3-Monooxygenase/KMO Polyclonal specifically detects Kynurenine 3-Monooxygenase/KMO in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Kynurenine 3-Monooxygenase/KMO
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1.02 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 ug/ml, ELISA 1:20000-1:60000, Immunohistochemistry 1:100-1:500, Immunocytochemistry/Immunofluorescence 1:100-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 50% (v/v) Glycerol with No Preservative
Gene Accession No. O15229
Gene Alias kynurenine 3-monooxygenase (kynurenine 3-hydroxylase)
Gene Symbols KMO
Host Species Rabbit
Immunogen Kynurenine 3-Monooxygenase/Kynurenine 3-Monooxygenase/KMO Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region surrounding 300-400aa of the mouse Kynurenine 3-Monooxygenase/KMO chain. (Uniprot: O15229)
Molecular Weight of Antigen 55.81 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Apoptosis, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 8564
Test Specificity This antibody is specific to KMO
Target Species Human, Mouse
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.