Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-161,982
GREENER_CHOICESDP

Catalog No. 251410 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-141-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 25-141-0 Supplier Tocris Bioscience™ Supplier No. 2514/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective EP4 receptor antagonist

L-161,982 is an EP4 receptor antagonist that is selective over all other members of the prostanoid receptor family (Ki values are 0.024, 0.71, 1.90, 5.10, 5.63, 6.74, 19 and 23 μM for human EP4, TP, EP3, DP, FP, IP, EP1 and EP2 receptors respectively). Suppresses PGE2-induced bone formation in rats and prevents the nociceptive response induced by Misoprostol (Cat. No. 2297) in formalin-injected mice.

Chemical Identifiers

CAS 147776-06-5
Molecular Formula C32H29F3N4O4S2

Specifications

Quantity 10 mg
Formula Weight 654.72
Purity >99%
Molecular Weight (g/mol) 654.723
Chemical Name or Material L-161, 982
Recommended Storage Desiccate at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.