Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-655,708
GSA_VAGREENER_CHOICE

Catalog No. 132710
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy

Catalog No. 13-271-0

Supplier: Tocris Bioscience™ 1327/10

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Benzodiazepine inverse agonist; selective for α5-containing GABAA receptors

Potent, selective inverse agonist for the benzodiazepine site of GABAA receptors containing the α5 subunit (Ki = 0.45 nM). Displays 50-100-fold selectivity over GABAA receptors containing α1, α2, α3 or α6 subunits in combination with β3 and γ2. Enhances LTP in a mouse hippocampal slice model and increases spatial learning, without displaying proconvulsant activity.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 130477-52-0
Molecular Formula C18H19N3O4

Specifications

Quantity 10 mg
Formula Weight 341.37
Purity >99%
Molecular Weight (g/mol) 341.367
Chemical Name or Material L-655, 708
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.