Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-685,458
GREENER_CHOICESDP

Catalog No. 26271 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
26-271 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 26-271 Supplier Tocris Bioscience™ Supplier No. 2627/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Potent and selective γ-secretase inhibitor

Potent and selective γ-secretase inhibitor (IC50 = 17 nM) that displays > 50-fold selectivity over a range of aspartyl, serine and cysteine proteases. Exhibits equal potency for inhibition of Aβ40 and Aβ42 peptides (IC50 values are 48 and 67 nM respectively in human neuroblastoma cells). Also regulates CXCR4 and VEGFR2 expression through inhibition of Notch signaling in vitro.

Chemical Identifiers

CAS 292632-98-5
Molecular Formula C39H52N4O6

Specifications

Quantity 1 mg
Formula Weight 672.85
Purity >97%
Molecular Weight (g/mol) 672.867
Chemical Name or Material L-685, 458
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.