Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-748,337
GSA_VAGREENER_CHOICE

Catalog No. 276010
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
27-601-0 10 mg
27-605-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 27-601-0 Supplier Tocris Bioscience™ Supplier No. 2760/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective β3 antagonist

Competitive β3-adrenoceptor antagonist that displays selectivity over β1 and β2 receptors (Ki values are 4.0, 204 and 390 nM for β3-, β2- and β1-adrenoceptors respectively). Inhibits cAMP accumulation in response to isoproterenol (IC50 = 6 nM). Reduces iNOS expression, attenuates nitric oxide-induced cell proliferation and induces apoptosis in a melanoma cell line.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Chemical Identifiers

CAS 244192-94-7
Molecular Formula C26H31N3O5S

Specifications

Quantity 10 mg
Formula Weight 497.61
Purity >98%
Molecular Weight (g/mol) 497.61
Chemical Name or Material L-748, 337
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.