Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L 760735
GREENER_CHOICE

Catalog No. 401310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-131-0 10 mg
40-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 40-131-0 Supplier Tocris Bioscience™ Supplier No. 4013/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

High affinity NK1 antagonist

High affinity NK1 receptor antagonist (IC50 = 0.19 nM at human NK1 receptors). Selective (>300-fold) over h-NK2 and h-NK3 receptors. Exhibits anxiolytic and antidepressant-like effects. Orally active.

Chemical Identifiers

CAS 188923-01-5
Molecular Formula C26H29ClF7N5O2

Specifications

Quantity 10 mg
Formula Weight 611.98
Purity >99%
Molecular Weight (g/mol) 611.99
Chemical Name or Material L 760735
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.