Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-AP4
GREENER_CHOICE

Catalog No. p-4783580
Click to view available options
Quantity:
1 mg
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Selective group III mGlu agonist

Selective group III metabotropic glutamate receptor agonist. Synaptic depressant. Agonist at the quisqualate-sensitized AP6 site in hippocampus.

Available as part of the Group III mGlu Receptor Tocriset™ and Mixed mGlu Receptor Tocriset™ . DL Mixture and D-isomer also available.

Chemical Identifiers

CAS 23052-81-5
Molecular Formula C4H10NO5P

Specifications

Quantity 1 mg
Formula Weight 183.1
Purity >99%
Molecular Weight (g/mol) 183.1
Chemical Name or Material L-AP4
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.