Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ L-AP5
GREENER_CHOICE

Catalog No. p-4783581
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective NMDA antagonist; less active form of DL-AP5

NMDA antagonist. Also agonist at quisqualate-sensitized AP6 site, where it is more potent than the isomer D-AP5 .

DL Mixture and D-isomer also available.

Chemical Identifiers

CAS 79055-67-7
Molecular Formula C5H12NO5P

Specifications

Quantity 10 mg
Formula Weight 197.13
Purity >99%
Molecular Weight (g/mol) 197.127
Chemical Name or Material L-AP5
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.