Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ L-Arabinose
SCI_ED

Catalog No. s25651 Shop All Fisherbrand Products
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy

Catalog No. S25651

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 5328-37-0
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key PYMYPHUHKUWMLA-VAYJURFESA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Density >1.0g/mL
Packaging Powder
Linear Formula C5H10O5
Quantity 25 g
Formula Weight 150.13
Chemical Name or Material L-Arabinose
ShelfLife 5 years
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.