Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-(+)-Arabinose, MP Biomedicals™

Catalog No. mp210070650 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 g
250 g

Used as a substrate to identify, differentiate and characterize pentose sugar isomerases. L-(+)-Arabinose, MP Biomedicals is the naturally occurring isomer and is a constituent of plant polysaccharides. It is used in the bioproduction of L-ribose

Most bacteria contain an inducible arabinose operon that codes for a series of enzymes and transporters that allows L-arabinose to be used as the sole carbon source in microbial culture.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5328-37-0
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13
InChI Key PYMYPHUHKUWMLA-VAYJURFESA-N
Synonym l-+-arabinose, dl-arabinose, 2r,3s,4s-2,3,4,5-tetrahydroxypentanal, aldehydo-l-arabinose, arabinose, l, unii-b40roo395z, pectinose, pectin sugar, aldehydo-l-arabino-pentose, arabinose,l
PubChem CID 5460291
ChEBI CHEBI:6182
IUPAC Name (2R,3S,4S)-2,3,4,5-tetrahydroxypentanal
SMILES C(C(C(C(C=O)O)O)O)O

Specifications

Melting Point 152°C to 165°C
Density 1.585g/mL (20°C) (Lit.)
Quantity 50 g
Solubility Information Freely soluble in water; Partially soluble in ethanol 90%.
Optical Rotation (c=1, water) 100–110°
Formula Weight 150.1
Percent Purity ≥98%
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.