Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ L-(+)-Ascorbic acid, 98+%

Manufacturer:  Alfa Aesar™ 11188A3

Quantity 2kg

 View more versions of this product

Catalog No. AA11188A3


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

50-81-7
176.12
84272
Vitamin C
C6H8O6
MFCD00064328
14,830
L-(+)-Ascorbic acid

Specifications

50-81-7
Powder
176.12
84272
Vitamin C
2kg
≥98%
1.65g/mL
C6H8O6
MFCD00064328
14,830
≈191°C (dec.)
L-(+)-Ascorbic acid

Safety and Handling

SDS & Safety Documents

RTECSNumber : CI7650000

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit