Learn More
L-(+)-Ascorbic acid, 99+%, Thermo Scientific Chemicals
Description
L-Ascorbic acid is an all important nutrient and antioxidant. L-ascorbic acid is frequently used as a processing aid or as a preservative in certain nutrients or food components. It is also utilized in the synthetic polymer industry, in photo processing and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents and in assay reagents.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
L-Ascorbic acid is an all important nutrient and antioxidant. L-ascorbic acid is frequently used as a processing aid or as a preservative in certain nutrients or food components. It is also utilized in the synthetic polymer industry, in photo processing and in metal technology. Miscellaneous uses have been proposed in cosmetics, fibers, preservation of blood, preservation of cut plants, cleaning agents and in assay reagents.
Solubility
Soluble in water. Insoluble in ether, chloroform, benzene, petroleum ether, oils, fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde.
Notes
Stable in air when dry.
Specifications
Melting Point | 190°C to 193°C (decomposition) |
Density | 1.65 |
Quantity | 50 g |
Beilstein | 84272 |
Merck Index | 14,830 |
Solubility Information | Soluble in water. Insoluble in ether,chloroform,benzene,petroleum ether,oils,fats and fat solvents. L-ascorbic acid means that the compound′s stereochemistry is related to that of the levorotatory enantiomer of glyceraldehyde. |
Optical Rotation | +21° (c=10 in Water) |
IUPAC Name | (5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxy-2,5-dihydrofuran-2-one |
Formula Weight | 176.12 |
Percent Purity | ≥99% |
Show More |
Safety and Handling
EINECSNumber | 200-066-2 |
RTECSNumber | CI7650000 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.