Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Cysteine, 99+%

Catalog No. p-3734470
Change view
Click to view available options
Quantity:
25 g
Packaging:
Plastic Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC173600250 25 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC173600250 Supplier Thermo Scientific Chemicals Supplier No. 173600250

L-Cysteine, CAS # 52-90-4, is an essential sulfur-containing amino acid related to cystine. It plays an important role in protein synthesis. It is a precursor to the glutathione antioxidant system and iron-sulfur clusters.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • L-Cysteine is an essential amino acid and is one of the building blocks required for the synthesis of proteins
  • This compound participates in the glutathione antioxidant system and iron-sulfur clusters

Applications

  • It plays a role in the production of flavors such as meat flavor obtained by reacting with sugars
  • This compound is also applied to the cleavage of disulfide bonds in keratin, the main protein in nails, skin, and hair
  • In chemical research, it is used as a crucial raw material in the manufacture of N-acetyl-L-cysteine, S-carboxymethyl-L-cysteine, and L-cysteine base
  • L-Cysteine is a component of the antioxidant glutathione and has been shown to be involved in the metabolism of essential biochemicals such as coenzyme A, heparin, and biotin
  • In vitro studies demonstrated that this compound can neutralize the activity of acetaminophen. In addition, due to its antioxidant activity this compound can protect the nervous system.
  • This compound can also bind to several metal cofactors in enzymes
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52-90-4
Molecular Formula C3H7NO2S
Molecular Weight (g/mol) 121.154
MDL Number MFCD00064306
InChI Key XUJNEKJLAYXESH-REOHCLBHSA-N
Synonym l-cysteine, cysteine, cystein, half-cystine, r-cysteine, thioserine, r-2-amino-3-mercaptopropanoic acid, l-+-cysteine, l-cystein, 2r-2-amino-3-sulfanylpropanoic acid
PubChem CID 5862
ChEBI CHEBI:17561
IUPAC Name (2R)-2-amino-3-sulfanylpropanoic acid
SMILES C(C(C(=O)O)N)S

Specifications

Ammonium 1000ppm max.
pH 4.5 to 6 (2.5% aq. soln.)
Color White
Chlorine (Cl) 400ppm max.
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Assay Percent Range 98+%
Loss on Drying 0.5% max. (105°C, 3 hrs)
Packaging Plastic Bottle
Linear Formula HSCH2CH(NH2)CO2H
Specific Rotation +8° to +9.5° (20°C, 589nm) (c=5, 2N HCl)
Sulfate 300ppm max.
Sulfated Ash 0.1% max.
Quantity 25 g
Formula Weight 121.15
Percent Purity ≥99.0% (on dried substance)
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L-Cysteine
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.