Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Glutamic Acid, FCC, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18603833 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 g
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

Catalog No. 18-603-833

Supplier: Spectrum Chemical Manufacturing Corporation G103650KGBL

Only null left
Add to Cart
Add to Cart

G1036, 56-86-0, C5H9NO4

Spectrum™ Chemical L-Glutamic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 0.1%
Packaging Fiber Drum
Quantity 50 kg
Optical Rotation +31.5° to +32.5°
Percent Purity 98.5 to 101.5%
Grade FCC
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.