Learn More
Description
L-Glutamic acid is a the predominant excitatory transmitter in the mammalian central nervous system. L-Glutamic acid acts at ionotropic and metabotropic glutamate receptors.
MNI-caged-L-glutamate (Cat. No. 1490) also available.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Quantity | 1 g |
| Product Line | Tocris |
| Formula Weight | 147.13 |
| Purity | >98% |
| Molecular Weight (g/mol) | 147.13 |
| Chemical Name or Material | L-Glutamic acid |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.


