Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Gulose 98.0+%, TCI America™
SDP

Catalog No. G023925G
Click to view available options
Quantity:
1 g
5 g
25 g

Chemical Identifiers

CAS 6027-89-0
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00136022
InChI Key GZCGUPFRVQAUEE-JGWLITMVSA-N
Synonym l-gulose, aldehydo-l-gulose, 2s,3s,4r,5s-2,3,4,5,6-pentahydroxyhexanal, unii-j96e9q45n7
PubChem CID 80127
ChEBI CHEBI:37701
IUPAC Name (2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O

Specifications

Melting Point 132°C
Color White
Quantity 25 g
Formula Weight 180.16
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material L-Gulose
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.