Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Iditol, 98+%, Thermo Scientific™

Catalog No. AC386541000
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 488-45-9
Molecular Formula C6H14O6
Molecular Weight (g/mol) 182.17
MDL Number MFCD00064289
InChI Key FBPFZTCFMRRESA-UHFFFAOYNA-N
Synonym l-iditol, 2s,3r,4r,5s-hexane-1,2,3,4,5,6-hexol, sorbo, l-idit
PubChem CID 5460044
ChEBI CHEBI:18202
IUPAC Name hexane-1,2,3,4,5,6-hexol
SMILES OCC(O)C(O)C(O)C(O)CO

Specifications

Melting Point 74.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Quantity 100 mg
Specific Rotation Condition − 24.00 (20.00°C c=1,water)
Specific Rotation − 24.00
Formula Weight 182.17
Percent Purity ≥98%
Physical Form Crystalline Powder
Chemical Name or Material L-Iditol, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.