Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ L-NMMA acetate

Click to view available options
Quantity:
10 mg
50 mg
Description
Competitive, irreversible inhibitor of all three NOS isoforms.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 248.28 |
| Purity | >98% |
| Molecular Weight (g/mol) | 248.283 |
| Chemical Name or Material | L-NMMA acetate |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction