Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ L-Photo-Leucine PROMO

Catalog No. PI22610PM
Click to view available options
:
Thermo Scientific™ L-Photo-Leucine

Diazirine analogs of leucine and methionine to express proteins in cell culture that will crosslink their protein interactors upon UV-light activation in vivo.

Thermo Scientific L-Photo-Leucine and L-Photo-Methionine are analogs of L-Leucine and L-Methionine amino acids that have activatable diazirine side chains capable of chemical crosslinking to adjacent molecules when exposed to ultraviolet light. Like their naturally occurring counterparts, these photo-reactive amino acids can be endogenously incorporated into the primary sequence of proteins during synthesis. Protein-protein interaction domains involving the synthesized proteins in their native environments can then be covalently crosslinked by activating the diazirine groups. Traditional crosslinking experiments require adding bifunctional chemical crosslinkers and associated reagent solvents, which can adversely affect the cell biology being studied. Using photo-reactive amino acids minimizes these potentially adverse effects and enables both stable and transient protein interactions in cells to be studied and characterized with a high degree of confidence.When used in combination with specially formulated limiting cell media that is devoid of leucine and methionine, the photo-activatable derivatives are treated like naturally occurring amino acids by the protein synthesis machinery. As a result, they can be substituted for leucine or methionine in the primary sequence of proteins. When exposed to UV light the diazirine rings become reactive intermediates that form covalent bonds with nearby protein side chains and backbones. Naturally associating binding partners are then instantly trapped. Crosslinked protein complexes are detected by decreased mobility on SDS-PAGE followed by Western blot detection, size-exclusion chromatography, sucrose density-gradient sedimentation or mass spectrometry.

Highlights:

  • In vivo labeling – incorporate photo-reactive group into proteins using normal cellular machinery

  • In vivo crosslinking – find interacting proteins in the native cellular environment

  • Increased specificity compared to traditional methods – crosslink interacting proteins correctly positioned at their interfaces within protein interaction domains

  • Complementary to traditional methods – provides detailed characterization of protein interactions when compared to results with formaldehyde and other more general crosslinking approaches

  • Efficient recovery – 90% protein recovery in cell lysates after crosslinking

  • Compatible – crosslink proteins expressed in a wide variety of cell lines, including HeLa, 293T, COS7, U2OS, A549, A431, HepG2, NIH 3T3 and C6

  • Easy to use – reagents are photo-stable under normal laboratory lighting, eliminating the need to work in the dark

Specifications

Chemical Reactivity Nonselective-NA (Metabolic labeling)
Cleavable No
Molecular Weight (g/mol) 143.15 g/mol
PEGylated No
Spacer Arm Length 0.0 Å
Content And Storage Upon receipt store at 4°C protected from light.
Cell Permeability Yes
Shipping Condition Ambient
Product Line Pierce™
Labeling Method Chemical Labeling, Metabolic Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Diazirine
Form Solid
Quantity 100 mg
Solubility Water
Format Standard
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.