Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L-Selectin/CD62L Antibody (95218), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB174131 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB174131 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB174131 Supplier Novus Biologicals Supplier No. FAB5761K
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

L-Selectin/CD62L Monoclonal antibody specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen L-Selectin/CD62L
Applications Flow Cytometry
Classification Monoclonal
Clone 95218
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse L-Selectin/CD62L, Met1-Asn332, Accession # P18337
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.5 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, B Cell Development and Differentiation Markers, Cardiovascular Biology, Glycobiology, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 6402
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.