Learn More
Description
Specifications
Specifications
| Antigen | L-Selectin/CD62L |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | DREG56 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen |
| Gene Alias | CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L |
| Gene Symbols | SELL |
| Host Species | Mouse |
| Immunogen | PMA-activated human peripheral blood leukocytes |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
