Learn More
L-Selectin/CD62L, Rabbit anti-Mouse, Clone: MEL-14, Novus Biologicals™
Shop All R&D Systems Products

Description
Specifications
Specifications
| Antigen | L-Selectin/CD62L |
| Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay |
| Classification | Monoclonal |
| Clone | MEL-14 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize |
| Formulation | PBS |
| Gene Alias | CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L |
| Gene Symbols | SELL |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.