Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
L-Selectin/CD62L, Rabbit anti-Mouse, Clone: MEL-14, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | L-Selectin/CD62L |
---|---|
Clone | MEL-14 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize |
Applications | Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay |
Description
L-Selectin/CD62L Monoclonal specifically detects L-Selectin/CD62L in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize.Specifications
L-Selectin/CD62L | |
1 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Blocking Assay | |
Unconjugated | |
Rabbit | |
Adaptive Immunity, B Cell Development and Differentiation Markers, Immunology, Lipid and Metabolism, Myeloid derived Suppressor Cell, Signal Transduction | |
PBS | |
6402 | |
C3H/eb mouse B lymphoma 38C-13. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
MEL-14 | |
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Block/Neutralize | |
Monoclonal | |
Purified | |
RUO | |
Mouse | |
CD62L, CD62L antigen, gp90-MEL, hLHRc, LAM-1, LAM1LECAM1, Leu-8, LEU8, Leukocyte adhesion molecule 1, Leukocyte surface antigen Leu-8, Leukocyte-endothelial cell adhesion molecule 1, LNHRTQ1, LSEL, L-selectin, Lyam-1, LYAM1CD62 antigen-like family member L, Lymph node homing receptor, lymphocyte adhesion molecule 1, pln homing receptor, PLNHR, selectin L | |
SELL | |
IgG κ | |
Protein A purified | |
This antibody binds to mouse L-Selectin/CD62L the mouse peripheral lymph node homing receptor, which is a lectin-like cell surface protein rapidly down-regulated upon cell activation with phorbol 12-myristate 13-acetate. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title