Learn More
Thermo Scientific Chemicals L-Thymidine
$137.70 - $423.00
Specifications
Format | Crystalline Powder |
---|---|
Molecular Weight (g/mol) | 242.23 |
Product Type | L-Thymidine |
Solution Clarity | (1% aq. soln.) Clear colorless solution |
Source | synthetic source |
Content And Storage | Refrigerator +4°C |
Shipping Condition | normal conditions |
Purity | ≥98% by HPLC |
Color | White to almost white |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AC464550010
|
Thermo Scientific Chemicals
464550010 |
1 g |
Each for $137.70
Order in Bulk
Request bulk or custom formats |
|
|||||
AC464550050
|
Thermo Scientific Chemicals
464550050 |
5 g |
Each for $423.00
Order in Bulk
Request bulk or custom formats |
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Crystalline Powder | |
L-Thymidine | |
synthetic source | |
normal conditions | |
White to almost white |
242.23 | |
(1% aq. soln.) Clear colorless solution | |
Refrigerator +4°C | |
≥98% by HPLC |
Safety and Handling
ShelfLife : 5 years
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.