Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ L-trans-2,4-PDC
GREENER_CHOICESDP

Catalog No. 029810 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-981-0 10 mg
02-985-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 02-981-0 Supplier R&D Systems™ Supplier No. 0298/10
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor

Potent, competitive, transportable EAAT1-4 inhibitor/non-transportable EAAT5 inhibitor. Also available as part of the Excitatory Amino Acid Transporter Inhibitor Tocriset™ .

Chemical Identifiers

CAS 64769-66-0
Molecular Formula C6H9NO4

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 159.14
Purity >97%
Molecular Weight (g/mol) 159.141
Chemical Name or Material L-trans-2, 4-PDC
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.