Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1/ORF2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177929 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177929 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177929 Supplier Novus Biologicals Supplier No. NBP177929
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

L1/ORF2 Polyclonal specifically detects L1/ORF2 in Human, Chimpanzee samples. It is validated for Western Blot, ELISA.

Specifications

Antigen L1/ORF2
Applications Western Blot, ELISA
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, ELISA 1:10000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q5BJF6
Gene Alias EC=2.7.7.49, EC=3.1.21.-, Endonuclease, L1, LINE-1 retrotransposable element ORF2 protein, Long interspersed element-1, ORF2p, Retrovirus-related Pol polyprotein LINE-1, Reverse transcriptase
Gene Symbols ODF2
Host Species Chicken
Immunogen This IgY fraction L1/ORF2 Antibody was prepared from eggs of chickens laid after repeated immunizations with two synthetic peptides conjugated to keyhole limpet hemocyanin (KLH). The peptides correspond to regions within the endonuclease domain of L1/ORF2 protein. (Uniprot: O00370)
Molecular Weight of Antigen 149.01 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4957
Test Specificity L1/ORF2 Antibody is directed against two regions within the endonuclease domain of L1 ORF2 protein. Reactivity occurs against human L1/ORF2 protein and is useful in determining its presence in various assays. A BLAST analysis was used to suggest cross reactivity with L1/ORF2 proteins from chimpanzee sources based on 100% homology with the immunizing sequences, and from macaque, fruit fly, cattle, dog, opossum, and rat sources based on 69 - 88% homology with the immunizing sequences.
Target Species Human, Chimpanzee
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.