Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1CAM Antibody (014), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB088395 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB088395 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB088395 Supplier Novus Biologicals Supplier No. NBP289294APC
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

L1CAM Monoclonal specifically detects L1CAM in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen L1CAM
Applications Flow Cytometry
Classification Monoclonal
Clone 14
Conjugate APC
Formulation PBS
Gene Alias antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human L1CAM (NP_000416.1; Met1-Glu1120).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Neuroscience, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 3897
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.