Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L1CAM Antibody (555) [Janelia Fluor™ 549], Novus Biologicals™
SDP

Catalog No. NB377729 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB377729 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB377729 Supplier Novus Biologicals Supplier No. FAB5674I
Add to Cart
Add to Cart

Rat Monoclonal Antibody

L1CAM Monoclonal antibody specifically detects L1CAM in Mouse samples. It is validated for Flow Cytometry, CyTOF

Specifications

Antigen L1CAM
Applications Flow Cytometry, CyTOF
Classification Monoclonal
Clone 555
Conjugate Janelia Fluor 549
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias antigen identified by monoclonal R1, CAML1N-CAML1, CD171, CD171 antigen, HSAS, HSAS1, L1 cell adhesion molecule, MASA, MIC5, N-CAM-L1, NCAM-L1, neural cell adhesion molecule L1, S10, SPG1
Host Species Rat
Immunogen Mouse cerebellum-derived partially purified L1CAM
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Glia Markers, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 3897
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.