Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ L1R Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144433
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144433 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144433 Supplier Invitrogen™ Supplier No. PA5144433
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

L1R Polyclonal Antibody for Western Blot, ELISA

Component of the entry fusion complex (EFC), which consists of 11 proteins. During cell infection, this complex mediates entry of the virion core into the host cytoplasm by a two-step mechanism consisting of lipid mixing of the viral and cellular membranes and subsequent pore formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen L1R
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene Alias Protein L1
Host Species Rabbit
Immunogen Recombinant Vaccinia virus Protein L1(L1R),partial (2-183aa).
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.