Learn More
Invitrogen™ L2HGDH Polyclonal Antibody

Description
L2HGDH Polyclonal Antibody for Western Blot
Specifications
Specifications
| Antigen | L2HGDH |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | L2HGDH |
| Gene Accession No. | Q91YP0, Q9H9P8 |
| Gene Alias | 2-hydroxyglutarate dehydrogenase; alpha-hydroxyglutarate oxidoreductase; alpha-ketoglutarate reductase; BC016226; C14orf160; Duranin; FLJ12618; L2HGA; L2HGDH; L-2-hydroxyglutarate dehydrogenase; L-2-hydroxyglutarate dehydrogenase, mitochondrial; L-alpha-hydroxyglutarate dehydrogenase; RGD1306250 |
| Gene Symbols | L2HGDH |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.