Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

L3HYPDH Antibody, Novus Biologicals™
SDP

Catalog No. NB287367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287367 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287367 Supplier Novus Biologicals Supplier No. NBP323479
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

L3HYPDH Polyclonal antibody specifically detects L3HYPDH in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen L3HYPDH
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS, pH 7.0
Gene Alias C14orf149, chromosome 14 open reading frame 149, EC 5.1.1.4, FLJ25436, probable proline racemase, proline racemase-like
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human L3HYPDH fragment
Purification Method Antigen and protein A Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112849
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.