Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
La-Related Protein 1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma ABE1800
Description
Anti-La-Related Protein 1 Antibody is a highly specific rabbit polyclonal antibody that targets La-related protein 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Specifications
La-Related Protein 1 | |
Polyclonal | |
Unconjugated | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human |
Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
Please refer to lot specific datasheet. | |
LARP1,KIAA0731,LARP | |
KLH-conjugated linear peptide corresponding to 15 amino acids from the N-terminal half of human La-related protein 1. | |
100 μg | |
Epigenetics and Nuclear Function | |
NP_056130 | |
Stable for 1 year at 2-8°C from date of receipt. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction