Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
La-Related Protein 1 Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $482.79
Specifications
Antigen | La-Related Protein 1 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABE1800-25UG
|
MilliporeSigma
ABE180025UG |
25 μg |
Each for $192.35
|
|
|||||
ABE1800MI
|
MilliporeSigma
ABE1800 |
100 μg |
Each for $482.79
|
|
|||||
Description
Anti-La-Related Protein 1 Antibody is a highly specific rabbit polyclonal antibody that targets La-related protein 1 and has been tested in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.Specifications
La-Related Protein 1 | |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. |
Please refer to lot specific datasheet. | |
Polyclonal | |
Rabbit | |
Epigenetics and Nuclear Function | |
LARP1,KIAA0731,LARP | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title