Learn More
Calprtectin-Monocyte/Macrophage/L1 protein Ab-1 Mouse Monoclonal Antibody, Epredia™
Shop All Epredia ProductsDescription
Host Species: Mouse
Clone: MAC 387
Isotype: IgG1
Species Reactivity: Human, Baboon, Monkey, Pig, Cat, Dog, Guinea pig, Rat, and Mouse. Others not known.
Epitope: Not determined
Immunogen: Affinity purified monocyte membrane preparation
Positive Control: Tonsil, lymph node, or spleen
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
Specifications
| Antigen | Macrophage/L1 Protein/Calprotectin Ab-1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | MAC 387 |
| Concentration | 200μg/mL |
| Conjugate | Unconjugated |
| Host Species | Mouse |
| Immunogen | Affinity purified monocyte membrane preparation |
| Quantity | 100 μL |
| Regulatory Status | IVD |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.