Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody

Test for macrophages and monocytes with Epredia™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody.

Supplier:  Epredia™ MS1808R7

 View more versions of this product

Catalog No. MS1808R7


Only null left
Add to Cart

Description

Description

CD68 glycoprotein is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: PG-M1

Isotype: IgG3/κ

Species Reactivity: Human. Others not tested

Epitope: Not determined

Immunogen: Gaucher cells

Molecular Weight: 110 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Specifications

Specifications

CD68/Macrophage Marker Ab-4
Monoclonal
Unconjugated
Gaucher cells
IVD
Primary
Prediluted
Immunohistochemistry (Paraffin)
PG-M1
Mouse
7 mL
Hematopoietic
Human
IgG3 κ
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.