Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody

Test for macrophages and monocytes with Epredia™ CD68/Macrophage Marker Ab-4, Mouse Monoclonal Antibody.

$205.00 - $944.00

Specifications

Antigen CD68/Macrophage Marker Ab-4
Clone PG-M1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 4
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Form Quantity Price Quantity & Availability  
MS1808S0
View Documents
Epredia™
MS1808S0
Concentrated 100 μL
Each for $205.00
Only null left
Add to Cart
 
MS1808S1
View Documents
Epredia™
MS1808S1
Concentrated 500 μL
Each for $686.00
Only null left
Add to Cart
 
MS1808S
View Documents
Epredia™
MS1808S
Concentrated 1 mL
Each for $944.00
Only null left
Add to Cart
 
MS1808R7
View Documents
Epredia™
MS1808R7
Prediluted 7 mL
Each for $500.00
Only null left
Add to Cart
 
Description

Description

CD68 glycoprotein is expressed on macrophages and monocytes.

Host Species: Mouse

Clone: PG-M1

Isotype: IgG3/κ

Species Reactivity: Human. Others not tested

Epitope: Not determined

Immunogen: Gaucher cells

Molecular Weight: 110 kDa

Positive Control: Tonsil

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

CD68/Macrophage Marker Ab-4
Immunohistochemistry (Paraffin)
Unconjugated
IVD
Human
IgG3 κ
PG-M1
Monoclonal
Mouse
Hematopoietic
Gaucher cells
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.