Learn More
Epredia™ Lab Vision™ CD79a/mb-1 (B-Cell Marker) Ab-2 Mouse Monoclonal Antibody, 200μg/mL, BSA and azide
Ensure accurate, reproducible resuts in immunohistochemistry applications with Epredia™ CD79a/mb-1 (B-Cell Marker) Ab-2, Mouse Monoclonal Antibody.
Specifications
| Antigen | CD79a/mb-1 (B-Cell Marker) Ab-2 |
|---|---|
| Clone | JCB117 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
|
MS1171P0
|
Epredia™
MS1171P0 |
Concentrated | 100 μL | N/A | N/A | N/A | |||
|
MS1171P1ABX
|
Epredia™
MS1171P1ABX |
Concentrated | 100 μL | N/A | N/A | N/A | |||
|
MS1171PABX
|
Epredia™
MS1171PABX |
Concentrated | 200 μL | N/A | N/A | N/A | |||
|
MS1171P1
|
Epredia™
MS1171P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
|
MS1171R1
|
Epredia™
MS1171R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
Description
A disulphide-linked heterodimer, consisting of mb-1 (or CD79a) and B29 (or CD79b) polypeptides, is non-covalently associated with membrane-bound immunoglobulins on B cells to constitute the B cell Ag receptor. CD79a first appears at pre B cell stage and persists until the plasma cell stage where it is found as an intracellular component. CD79a is found in the majority of acute leukemias of precursor B cell type, in B cell lines, B cell lymphomas, and in some myelomas. It is not present in myeloid or T cell lines.Host Species: Mouse
Clone: JCB117
Isotype: IgG1/κ
Species Reactivity: Human. Others not known.
Epitope: Extracellular domain
Immunogen: Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein
Molecular Weight: 50kDa
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
| CD79a/mb-1 (B-Cell Marker) Ab-2 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| IVD | |
| Human | |
| IgG2a κ |
| JCB117 | |
| Monoclonal | |
| Mouse | |
| B-Cell | |
| Recombinant protein containing part of the extracellular portion of the CD79a glycoprotein | |
| Primary |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.