Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ COX2 Rabbit Polyclonal Antibody with BSA and azide

Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Epredia™ COX2, Rabbit Polyclonal Antibody.

Supplier:  Epredia™ RB9072P

 View more versions of this product

Catalog No. RB9072P

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

COX2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate-early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse, and Rat. Others not tested.

Epitope: C-terminus

Immunogen: Synthetic peptide corresponding to C-terminus of rat COX2

Molecular Weight: 70 kDa

Positive Control: HT29 cells, Lung and colon CA

Cellular Localization: Cytoplasmic and membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

COX2
Polyclonal
200µg/mL
Rabbit
1 mL
Enzyme
Human, Mouse
IgG
Immunoprecipitation, Western Blot
Polyclonal
Unconjugated
Synthetic peptide corresponding to c-terminus of rat COX2
IVD
Primary
Concentrated
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.