Learn More
Epredia™ Lab Vision™ COX2 Rabbit Polyclonal Antibody with BSA and azide
Ensure accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments with Epredia™ COX2, Rabbit Polyclonal Antibody.
Specifications
Antigen | COX2 |
---|---|
Applications | Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Quantity | Price | Quantity & Availability | ||||
RB9072P1
|
Epredia™
RB9072P1 |
Concentrated | 500 μL | N/A | N/A | N/A | |||
RB9072P
|
Epredia™
RB9072P |
Concentrated | 1 mL | N/A | N/A | N/A | |||
RB9072R1
|
Epredia™
RB9072R1 |
Prediluted | 1 mL | N/A | N/A | N/A | |||
RB9072R7
|
Epredia™
RB9072R7 |
Prediluted | 7 mL | N/A | N/A | N/A | |||
Description
COX2 (Cyclooxygenase-2) is an inducible enzyme. It is involved in the response of cells to growth factors, tumor promoters, and cytokines that induce its expression. Given its role in synthesizing prostaglandins, COX2 is therefore of interest in studying immune response regulation. COX2 is induced by a wide variety of stimuli and was initially identified as immediate-early growth response gene. In addition, COX2 expression markedly increased in 85-90% of human colorectal adenocarcinoma whereas COX1 levels remain unchanged.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse, and Rat. Others not tested.
Epitope: C-terminus
Immunogen: Synthetic peptide corresponding to C-terminus of rat COX2
Molecular Weight: 70 kDa
Positive Control: HT29 cells, Lung and colon CA
Cellular Localization: Cytoplasmic and membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
COX2 | |
Polyclonal | |
Rabbit | |
Enzyme | |
Synthetic peptide corresponding to c-terminus of rat COX2 | |
Primary |
Immunoprecipitation, Western Blot | |
Unconjugated | |
IVD | |
Human, Mouse | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.