Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Epredia™ Lab Vision™ Cytokeratin 18 Rabbit Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry experiments with Epredia™ Cytokeratin 18, Rabbit Monoclonal Antibody.

$6752.00

Specifications

Antigen Cytokeratin 18
Clone E431-1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Form Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Form Price Quantity & Availability  
RM2108S
View Documents
Epredia™
RM2108S
1 mL Concentrated
Each for $6,752.00
Only null left
Add to Cart
 
RM2108S0
View Documents
Epredia™
RM2108S0
100 μL Concentrated N/A N/A N/A
RM2108R1
View Documents
Epredia™
RM2108R1
1 mL Prediluted N/A N/A N/A
RM2108R7
View Documents
Epredia™
RM2108R7
7 mL Prediluted N/A N/A N/A
RM2108RQ
View Documents
Epredia™
RM2108RQ
12 mL Ready-to-Use N/A N/A N/A
Description

Description

Cytokeratin 18, which belongs to the type A (acidic) subfamily of low molecular weight keratins, exists in combination with cytokeratin 8. Tissues from gastrointestinal tract, respiratory tract and urogenital tract, as well as endocrine and exocrine tissues and mesothelial cells are positive for cytokeratin 18. Typically, it is absent in squamous cell carcinoma.

Host Species: Rabbit

Clone: E431-1

Isotype: IgG

Species Reactivity: Human. Others not known.

Immunogen: A synthetic peptide corresponding to residues of the human CK18

Molecular Weight: 45 kDa

Positive Control: Mesothelioma, Pancreas

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)

Specifications

Specifications

Cytokeratin 18
Immunohistochemistry (Paraffin)
Unconjugated
IVD
A synthetic peptide corresponding to residues of the human CK18
E431-1
Monoclonal
Rabbit
Cancer and Tumor Biology
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.