Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Desmin (Muscle Cell Marker) Rabbit Polyclonal Antibody, Epredia™

Obtain accurate, reproducible results in immunohistochemistry, western blotting and immunoprecipitation experiments.

Supplier:  Epredia RB9014P0

 View more versions of this product

Catalog No. RB9014P0


Description

Description

Desmin is an intermediate filament protein of both smooth and striated muscles. Antibody to desmin reacts with striated (skeletal and cardiac) as well as smooth muscle cells. In skeletal and cardiac muscles, the staining is confined to the Z-bands giving a characteristic striated appearance. Anti-desmin antibody is useful in identification of tumors of myogenic origin.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human, Mouse and Rat. Others not tested.

Epitope: Near C-terminus

Immunogen: A synthetic peptide mapping near the C-terminus of human desmin

Molecular Weight: 53 kDa

Positive Control: Muscle or Sarcoma

Cellular Localization: Cytoplasmic

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Immunoprecipitation
Specifications

Specifications

Desmin (Muscle Cell Marker)
Polyclonal
Unconjugated
A synthetic peptide mapping near the C-terminus of human desmin
IVD
Primary
Concentrated
Immunoprecipitation, Western Blot
200µg/mL
Rabbit
100 μL
Muscle Biology
Human, Mouse
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.