Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Mouse and Rat. Others not tested.
Epitope: Near C-terminus
Immunogen: A synthetic peptide mapping near the C-terminus of human desmin
Molecular Weight: 53 kDa
Positive Control: Muscle or Sarcoma
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation
Specifications
Specifications
| Antigen | Desmin (Muscle Cell Marker) |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 200µg/mL |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide mapping near the C-terminus of human desmin |
| Quantity | 500 μL |
| Regulatory Status | IVD |
| Research Discipline | Muscle Biology |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.